Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fetuin Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299415JF646
Description
Fetuin Polyclonal specifically detects Fetuin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Fetuin | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
fetuin B, fetuin-B, Fetuin-like protein IRL685,16G2, Gugu, IRL685 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human Fetuin. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
26998 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction