Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fetuin B Antibody (019), Alexa Fluor™ 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290642AF350
Description
Fetuin B Monoclonal antibody specifically detects Fetuin B in Mouse samples. It is validated for ELISA.Specifications
Fetuin B | |
Monoclonal | |
Alexa Fluor 350 | |
FETUB, fetuin-B, Fetuin-like protein IRL685,16G2, Gugu, IRL685 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Fetuin B (Q9QXC1-1; Met 1-Pro 388). | |
0.1 mL | |
Primary | |
Has cross-reactivity in ELISA with: Human FETUB | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
19 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
26998 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction