Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fetuin B Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298307MFV450
Description
Fetuin B Polyclonal antibody specifically detects Fetuin B in Mouse samples. It is validated for ELISASpecifications
Fetuin B | |
Polyclonal | |
ELISA | |
FETUB, fetuin beta;fetuin phosphoprotein, fetuin-B, Fetuin-like protein IRL685,16G2, Gugu, IRL685 | |
Produced in rabbits immunized with purified, recombinant Mouse Fetuin B (Uniprot#: Q9QXC1-1; Met 1-Pro 388) | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
ELISA | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
26998 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction