Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fetuin B Polyclonal Antibody

Catalog No. PIPA5121997
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121997 20 μL
PIPA5119113 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121997 Supplier Invitrogen™ Supplier No. PA5121997
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Fetuin B Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

The protein encoded by this gene is a member of the fetuin family, part of the cystatin superfamily of cysteine protease inhibitors. Fetuins have been implicated in several diverse functions, including osteogenesis and bone resorption, regulation of the insulin and hepatocyte growth factor receptors, and response to systemic inflammation. This protein may be secreted by cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fetuin B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene FETUB
Gene Accession No. Q9QX79, Q9QXC1, Q9UGM5
Gene Alias 16G2; 2310011O17Rik; AI255764; D17980; Fet; FETUB; fetuin B; fetuin beta; fetuin phosphoprotein; fetuin-B; Fetuin-like protein IRL685; Gugu; IRL685; Pp63
Gene Symbols FETUB
Host Species Rabbit
Immunogen Recombinant protein Fetuin B. The antigen corresponds to amino acid range 152-261 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26998, 59083, 83928
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.