Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Fetuin, From Fetal Bovine Serum
Click to view available options
Quantity:
100 mg
250 mg
1 g
Description
- Formulation: Polypeptide 74%, Hexose 8.3%, Hexosamines 5.5%, and Sialic Acid 8.7%
Applications:
Fetuin bovine is a Protein with the ability to bind to the insulin receptor and inhibit insulin action, Fetuin has been used in studies of type 2 diabetes mellitus (T2DM), In patients with T2DM, fetuin has been shown to be significantly lower when compared to control patients.

Specifications
Specifications
For Use With (Application) | Type 2 Diabetes Mellitus (T2DM) Studies, Cell Attachment and Proliferation |
Formulation | Polypeptide 74%, Hexose 8.3%, Hexosamines 5.5%, and Sialic Acid 8.7% |
Name | Fetuin Protein |
Purification Method | Ammonium Sulfate Fractionation |
Quantity | 1 g |
Source | Fetal Bovine Serum |
Form | Powder |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction