Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fetuin Rabbit anti-Mouse, Clone: 110, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | Fetuin |
---|---|
Clone | 110 |
Dilution | ELISA 1:1000-1:2000 |
Applications | ELISA |
Classification | Monoclonal |
Description
Fetuin Monoclonal specifically detects Fetuin in Mouse samples. It is validated for ELISA.Specifications
Fetuin | |
ELISA 1:1000-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
26998 | |
IgG | |
Protein A purified |
110 | |
ELISA | |
Unconjugated | |
Rabbit | |
Mouse | |
fetuin B, fetuin-B, Fetuin-like protein IRL685,16G2, Gugu, IRL685 | |
Recombinant Mouse Fetuin protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title