Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FetuinGoat anti-Human, Polyclonal, Rockland™
SDP

Catalog No. 501990325
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0325 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-0325 Supplier Rockland Immunochemicals Supplier No. 200101456
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Goat Serum.

Alpha2-HS glycoprotein (AHSG), a glycoprotein present in the serum, is synthesized by hepatocytes. The AHSG molecule consists of two polypeptide chains, which are both cleaved from a proprotein encoded from a single mRNA. It is involved in several functions, such as endocytosis, brain development and the formation of bone tissue. The protein is commonly present in the cortical plate of the immature cerebral cortex and bone marrow hemopoietic matrix, and it has therefore been postulated that it participates in the development of the tissues. However, its exact significance is still obscure.

Specifications

Antigen Fetuin
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene AHSG
Gene Accession No. P02765
Gene Alias 59 kDa bone sialic acid-containing protein; A2HS; Aa2-066; AHS; Ahsg; alpha 2 HS-glycoprotein alpha 2 (fetuin); alpha 2-HS glycoprotein; Alpha-2-HS-glycoprotein; Alpha-2-HS-glycoprotein chain A; Alpha-2-HS-glycoprotein chain B; Alpha-2-Z-globulin; Asialofetuin; Ba-alpha-2-glycoprotein; BOS_906; BSP; Countertrypin; FETUA; fetuin A; FetuinA; Fetuin-A; Glycoprotein PP63; HSGA; phosphorylated N-glycoprotein pp63; pp63; PRO2743
Gene Symbols AHSG
Host Species Goat
Immunogen This purified antibody was prepared from rabbit serum after repeated immunizations with a recombinant human fetuin (a2-HS glycoprotein) processed to remove a 40 amino acid residue bridging peptide resulting in the mature form of the protein.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 197
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.