Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FEV Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598805
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598805 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598805 Supplier Invitrogen™ Supplier No. PA598805
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FEV Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

This gene belongs to the ETS transcription factor family. ETS family members have a highly conserved 85-amino acid ETS domain that binds purine-rich DNA sequences. The alanine-rich C-terminus of this gene indicates that it may act as a transcription repressor. This gene is exclusively expressed in neurons of the central serotonin (5-HT) system, a system implicated in the pathogeny of such psychiatric diseases as depression, anxiety, and eating disorders. In some types of Ewing tumors, this gene is fused to the Ewing sarcoma (EWS) gene following chromosome translocations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FEV
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene FEV
Gene Accession No. O70132, Q99581
Gene Alias ETS domain transcription factor Pet-1; ETS-domain transcription factor Pet-1; Fev; FEV (ETS oncogene family); FEV (fifth Ewing variant); FEV transcription factor, ETS family member; FEV, ETS transcription factor; fifth Ewing variant protein; HSRNAFEV; mPet-1; PC12 ETS domain-containing transcription factor 1; PC12 ETS factor 1; Pet1; Pet-1; pheochromocytoma 12 ETS (E26 transformation-specific); protein FEV
Gene Symbols FEV
Host Species Rabbit
Immunogen Recombinant Human Protein FEV protein (1-97AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246271, 54738
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.