Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FEZ1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520396
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520396 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520396 Supplier Invitrogen™ Supplier No. PA520396
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Despite its predicted molecular weight, FEZ1 often migrates at a higher molecular weight in SDS-PAGE. A suggested positive control is SK-N-SH cell lysate. PA5-20396 can be used with blocking peptide PEP-0513.

This gene is an ortholog of the C. elegans unc-76 gene, which is necessary for normal axonal bundling and elongation within axon bundles. Expression of this gene in C. elegans unc-76 mutants can restore to the mutants partial locomotion and axonal fasciculation, suggesting that it also functions in axonal outgrowth. The N-terminal half of the gene product is highly acidic. Alternatively spliced transcript variants encoding different isoforms of this protein have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FEZ1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Fez1
Gene Accession No. P97577, Q8K0X8, Q99689
Gene Alias F37; fasciculation and elongation protein zeta 1; fasciculation and elongation protein zeta 1 (zygin I); fasciculation and elongation protein zeta-1; Fez1; protein kinase C-binding protein Zeta1; UNC76; UNC-76; zygin 1; zygin I; Zygin-1
Gene Symbols Fez1
Host Species Rabbit
Immunogen A 19 amino acid peptide from near the carboxy terminus of human FEZ1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 235180, 81730, 9638
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.