Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FEZ1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | FEZ1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
FEZ1 Polyclonal specifically detects FEZ1 in Human samples. It is validated for Western Blot.Specifications
FEZ1 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
fasciculation and elongation protein zeta 1 (zygin I), fasciculation and elongation protein zeta-1, Zygin I, zygin-1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human FEZ1 (NP_005094.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Tumor Suppressors | |
PBS with 50% glycerol, pH7.3. | |
9638 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title