Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FEZ2 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281716V
This item is not returnable.
View return policy
Description
FEZ2 Polyclonal antibody specifically detects FEZ2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
FEZ2 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA, Immunofluorescence | |
DyLight 405 | |
fasciculation and elongation protein zeta 2 (zygin II) | |
FEZ2 antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human FEZ2. The immunogen is located within the last 50 amino acids of FEZ2. Amino Acid Squence: IPYEKKNGPPSVEDL | |
0.1 mL | |
Neuroscience | |
9637 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction