Learn More
Invitrogen™ FEZ2 Polyclonal Antibody

Description
A suggested positive control is mouse brain tissue lysate. PA5-20456 can be used with blocking peptide PEP-0573.
Specifications
Specifications
| Antigen | FEZ2 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | FEZ2 |
| Gene Accession No. | P97578, Q6TYB5, Q9UHY8 |
| Gene Alias | 9030616F16; D17Ertd315e; fasciculation and elongation protein zeta 2; fasciculation and elongation protein zeta 2 (zygin II); fasciculation and elongation protein zeta-2; FEZ2; HUM3CL; pre-T/NK cell associated protein (3Cl); synaptotagmin interacting protein zyginII; Zrp; zygin 2; zygin II; Zygin-2; zygin-related protein types I/II |
| Gene Symbols | FEZ2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.