Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FEZ2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119105
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119105 200 μL
PIPA5121996 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119105 Supplier Invitrogen™ Supplier No. PA5119105
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FEZ2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Fasciculation and elongation protein zeta-2 (FEZ2) is a homolog to the mammalian FEZ1, itself an ortholog of the C. elegans UNC-76. In contrast to FEZ1, FEZ2 mRNA is widely expressed in mouse tissues. FEZ2 interacts with protein kinase C (PKC)-zeta substrate and induces neurite extension of PC12 cells when co-expressed with a constitutively active form of PKC-zeta, suggesting FEZ2 may play an important role in the morphological changes of various cells by associating with PKC-zeta in a tissue non-specific manner. FEZ2 can interact with FEZ1 through its c-terminal regions and especially its coiled-coil region. At least two isoforms of FEZ2 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FEZ2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene FEZ2
Gene Accession No. P97578, Q9UHY8
Gene Alias 9030616F16; D17Ertd315e; fasciculation and elongation protein zeta 2; fasciculation and elongation protein zeta 2 (zygin II); fasciculation and elongation protein zeta-2; FEZ2; HUM3CL; pre-T/NK cell associated protein (3Cl); synaptotagmin interacting protein zyginII; Zrp; zygin 2; zygin II; Zygin-2; zygin-related protein types I/II
Gene Symbols FEZ2
Host Species Rabbit
Immunogen Recombinant protein Fasciculation And Elongation Protein Zeta 2. The antigen corresponds to amino acid range 1-375 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94269, 9637
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.