Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FEZF1 Polyclonal Antibody

Catalog No. PIPA566700
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566700 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566700 Supplier Invitrogen™ Supplier No. PA566700

Rabbit Polyclonal Antibody

Immunogen sequence: YLAERNKLVVP AVEKYPSGVA FKDLSQAQLQ HYMKESAQLL SEKIAFKTSD FSRGSPNAKP KV Highest antigen sequence identity to the following orthologs - mouse 98%, rat 95%.

Transcription repressor. Involved in the axonal projection and proper termination of olfactory sensory neurons (OSN). Plays a role in rostro-caudal patterning of the diencephalon and in prethalamic formation. Expression is required in OSN to cell-autonomously regulate OSN axon projections. Regulates non-cell-autonomously the layer formation of the olfactory bulb development and the interneurons. May be required for correct rostral migration of the interneuron progenitors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FEZF1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FEZF1
Gene Accession No. A0PJY2
Gene Alias 3110069A13Rik; FEZ; Fez family zinc finger 1; fez family zinc finger protein 1; Fezf1; fez-like; HH22; LOW QUALITY PROTEIN: fez family zinc finger protein 1; RGD1560480; Zfp312-like; zinc finger protein 312B; ZNF312B
Gene Symbols FEZF1
Host Species Rabbit
Immunogen Recombinant Human FEZF1. Recombinant protein control fragment (Product #RP-107351).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389549
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.