Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FFAR1/GPR40 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB1001537
Description
FFAR1/GPR40 Polyclonal specifically detects FFAR1/GPR40 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
FFAR1/GPR40 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
free fatty acid receptor 1, G protein-coupled receptor 40, GPCR40, GPR40FFA1R, G-protein coupled receptor 40 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog and Cow. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 0.01 - 0.03 ug/ml, Peptide ELISA Detection limit 1:16000 | |
NP_005294.1 | |
FFAR1 | |
Peptide with sequence CAARTQGGKSQK corresponding to C-Terminus according to NP_005294.1. | |
0.1 mg | |
GPCR | |
2864 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction