Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FFAR2/GPR43 Antibody (1000624) [DyLight 550], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB10082L
Description
FFAR2/GPR43 Monoclonal antibody specifically detects FFAR2/GPR43 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
FFAR2/GPR43 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
1000624 | |
Flow Cytometry, CyTOF-ready | |
free fatty acid activated receptor 2, free fatty acid receptor 2, G protein-coupled receptor 43, GPR43FFA2R, G-protein coupled receptor 43 | |
NS0 Mouse myeloma cell line transfected with human FFAR2/GPR43, Met1-Glu330, Accession # O15552 | |
0.1 mL | |
GPCR | |
2867 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction