Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FFAR3/GPR41 Antibody (2590G) [Allophycocyanin], Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB10562A
Description
FFAR3/GPR41 Monoclonal antibody specifically detects FFAR3/GPR41 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
FFAR3/GPR41 | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
2590G | |
Western Blot, Flow Cytometry, CyTOF-ready | |
free fatty acid receptor 3, G protein-coupled receptor 41, GPR41FFA3R, G-protein coupled receptor 41 | |
Synthetic peptide containing human FFAR3 | |
0.1 mL | |
GPCR | |
2865 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction