Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FFAR3/GPR41 Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP303618100UL
Description
FFAR3/GPR41 Polyclonal specifically detects FFAR3/GPR41 in Rat samples. It is validated for Western Blot.Specifications
FFAR3/GPR41 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Rat | |
Purified |
Western Blot | |
Unconjugated | |
free fatty acid receptor 3, G protein-coupled receptor 41, GPR41FFA3R, G-protein coupled receptor 41 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 257-346 of human FFAR3/GPR41 (NP_005295.1). | |
100 μL | |
GPCR | |
2865 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction