Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FFAR3/GPR41 Rabbit anti-Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB095550 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB095550 20 μL
NB095549 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB095550 Supplier Novus Biologicals Supplier No. NBP30361820UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FFAR3/GPR41 Polyclonal specifically detects FFAR3/GPR41 in Rat samples. It is validated for Western Blot.

Specifications

Antigen FFAR3/GPR41
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias free fatty acid receptor 3, G protein-coupled receptor 41, GPR41FFA3R, G-protein coupled receptor 41
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 257-346 of human FFAR3/GPR41 (NP_005295.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 2865
Target Species Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.