Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FFAR3 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc FFAR3301AP

Description
FFAR3 Polyclonal antibody specifically detects FFAR3 in Human, Mouse, Rat samples. It is validated for ELISA, Immunoprecipitation, Western BlotSpecifications
FFAR3 | |
Polyclonal | |
Unconjugated | |
FFAR3 | |
FFA3R; FFAR3; Free fatty acid receptor 3; G protein-coupled receptor 41; Gm478; Gpcr41; GPR41; GPR42; G-protein coupled receptor 41; putative G protein-coupled receptor GPR41 | |
Synthetic peptide taken within amino acid region 270-320 on human FFAR3 protein. | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
B2GV46, O14843, Q3UFD7 | |
Rabbit | |
Affinity chromatography | |
RUO | |
233080, 2865, 365228 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction