Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FFAR4/GPR120 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP100858AF700
Description
FFAR4/GPR120 Polyclonal specifically detects FFAR4/GPR120 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
FFAR4/GPR120 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated | |
G protein-coupled receptor 120, G protein-coupled receptor 129, G protein-coupled receptor PGR4, GPR120, GPR129, G-protein coupled receptor 120, G-protein coupled receptor 129, G-protein coupled receptor GT01, G-protein coupled receptor PGR4, G-protein-coupled receptor GT01, GT01, MGC119984, omega-3 fatty acid receptor 1, PGR4DKFZp686F0824 | |
Rabbit | |
0.1 mL | |
338557 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay | |
Alexa Fluor 700 | |
50 mM sodium borate with 0.05% sodium azide | |
FFAR4 | |
A synthetic peptide made to an internal portion of the human GPR120 protein (between residues 200-300) [UniProt Q5NUL3] | |
Primary | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction