Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FFAR4/GPR120 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP189739
Description
FFAR4/GPR120 Polyclonal specifically detects FFAR4/GPR120 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FFAR4/GPR120 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:10 - 1:20 | |
Q5NUL3 | |
FFAR4 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:FRVVPQRLPGADQEISICTLIWPTIPGEISWDV | |
0.1 mL | |
GPCR | |
338557 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
G protein-coupled receptor 120, G protein-coupled receptor 129, G protein-coupled receptor PGR4, GPR120, GPR129, G-protein coupled receptor 120, G-protein coupled receptor 129, G-protein coupled receptor GT01, G-protein coupled receptor PGR4, G-protein-coupled receptor GT01, GT01, MGC119984, omega-3 fatty acid receptor 1, PGR4DKFZp686F0824 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction