Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ FFN 200 dihydrochloride
GREENER_CHOICESDP

Catalog No. 591110 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-111-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 59-111-0 Supplier R&D Systems™ Supplier No. 5911/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective fluorescent VMAT2 substrate

FFN 200 dihydrochloride is a selective fluorescent VMAT2 substrate. Selectively traces exocytosis in dopaminergic neuronal cell culture and brain tissue. Excitation and emission maxima are 352 and 451 nm, respectively.

Specifications

Chemical Name or Material 4-(2-Aminoethyl)-7-amino-2H-1-benzopyran-2-one dihydrochloride
CAS 2080306-27-8
Quantity 10 mg
Solubility Soluble to 100mM in water and to 100mM in DMSO
Product Line Tocris
Target Fluorescent Transporter Probes
Molecular Formula C11H12N2O20.2 HCl
Formula Weight 277.15
Physical Form Solid
Color White
Product Type Bioactive Small Molecule
Purity >98%
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.