Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ FFN 270
GREENER_CHOICESDP

Catalog No. 671710 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-171-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 67-171-0 Supplier R&D Systems™ Supplier No. 6717/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Fluorescent substrate for NET and VMAT2

FFN 270 is a fluorescent false neurotransmitter (FFN). Fluorescent substrate for NET and VMAT2. Labels noradrenergic neurons and their synaptic vesicles, and enables imaging of synaptic vesicle content release from specific axonal sites in vivo. Selectively labels NA neurons over other monoamine and CNS targets. Exhibits two resolved absorption/excitation maxima depending on solvent pH (excitation maxima: 320 nm or 365 nm, emission maxima: 475 nm).

Specifications

Chemical Name or Material 3-(2-Aminoethyl)-8-fluoro-7-hydroxy-2H-chromen-2-one hydrochloride
CAS 2341841-05-0
Quantity 10 mg
Solubility Soluble to 100mM in DMSO
Product Line Tocris
Target Fluorescent Transporter Probes
Molecular Formula C11H10FNO3.HCl
Formula Weight 259.66
Physical Form Solid
Color Beige
Product Type Fluorescent substrate
Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.