Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FGD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541477
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541477 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541477 Supplier Invitrogen™ Supplier No. PA541477
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: HGHRAPGGAG PSEPEHPATN PPGAAPPACA DSDPGASEPG LLARRGSGSA Sequence homology: Cow: 86%; Human: 100%; Pig: 86%; Rat: 79%.

FGD1 contains Dbl (DH) and pleckstrin (PH) homology domains. It can bind specifically to the Rho family GTPase Cdc42Hs and stimulate the GDP-GTP exchange of the isoprenylated form of Cdc42Hs. It also stimulates the mitogen activated protein kinase cascade leading to c-Jun kinase SAPK/JNK1 activation. FGD1 has an essential role in embryonic development, and FGD1 gene mutations result in the human developmental disorder, Aarskog-Scott syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGD1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FGD1
Gene Accession No. P98174
Gene Alias AAS; faciogenital dysplasia 1 protein; Faciogenital dysplasia 1 protein homolog; faciogenital dysplasia protein; Fgd1; FGDY; FYVE, RhoGEF and PH domain containing 1; FYVE, RhoGEF and PH domain containing 1 (faciogenital dysplasia); FYVE, RhoGEF and PH domain-containing protein 1; MRXS16; Rho/Rac GEF; rho/Rac guanine nucleotide exchange factor FGD1; ZFYVE3; zinc finger FYVE domain-containing protein 3
Gene Symbols FGD1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human FGD1 (aa 2-51).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2245
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.