Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FGD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551482
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551482 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551482 Supplier Invitrogen™ Supplier No. PA551482
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSDPGASEPG LLARRGSGSA LGGPLDPQFV GPSDTSLGAA PGHRVLPCGP SPQHHRALRF SYHLEGSQPR PGLHQGNRIL VKSLSLDPGQ SLEPHPEGPQ RLRSDP Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 31%.

FGD1 contains Dbl (DH) and pleckstrin (PH) homology domains. It can bind specifically to the Rho family GTPase Cdc42Hs and stimulate the GDP-GTP exchange of the isoprenylated form of Cdc42Hs. It also stimulates the mitogen activated protein kinase cascade leading to c-Jun kinase SAPK/JNK1 activation. FGD1 has an essential role in embryonic development, and FGD1 gene mutations result in the human developmental disorder, Aarskog-Scott syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGD1
Applications Western Blot, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FGD1
Gene Accession No. P98174
Gene Alias AAS; faciogenital dysplasia 1 protein; Faciogenital dysplasia 1 protein homolog; faciogenital dysplasia protein; Fgd1; FGDY; FYVE, RhoGEF and PH domain containing 1; FYVE, RhoGEF and PH domain containing 1 (faciogenital dysplasia); FYVE, RhoGEF and PH domain-containing protein 1; MRXS16; Rho/Rac GEF; rho/Rac guanine nucleotide exchange factor FGD1; ZFYVE3; zinc finger FYVE domain-containing protein 3
Gene Symbols FGD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FGD1. Recombinant protein control fragment (Product #RP-91933).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2245
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.