Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30396620UL
Description
FGD1 Polyclonal specifically detects FGD1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
FGD1 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
2245 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
faciogenital dysplasia (Aarskog-ScRhoGEF and PH domain-containing protein 1, Faciogenital dysplasia 1 protein, FGDYMRXS16, FYVE, RhoGEF and PH domain containing 1, Rho/Rac guanine nucleotide exchange factor FGD1, ZFYVE3AAS, Zinc finger FYVE domain-containing protein 3 | |
A synthetic peptide corresponding to a sequence within amino acids 700-800 of human FGD1 (NP_004454.2). | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction