Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FGD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548526
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548526 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548526 Supplier Invitrogen™ Supplier No. PA548526
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FGD2 Polyclonal Antibody for Western Blot, IHC (P)

Activates CDC42, a member of the Ras-like family of Rho- and Rac proteins, by exchanging bound GDP for free GTP. Activates JNK1 via CDC42 but not RAC1. Binds to phosphatidylinositol 4,5- bisphosphate, phosphatidylinositol 3,4,5-trisphosphate, phosphatidylinositol 5-monophosphate, phosphatidylinositol 4- monophosphate and phosphatidylinositol 3-monophosphate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGD2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene FGD2
Gene Accession No. Q7Z6J4, Q8BY35
Gene Alias faciogenital dysplasia homolog 2; faciogenital dysplasia protein 2; Fgd1 family, member 2; Fgd2; Fgd2-like protein; FLJ00276 protein; FYVE, RhoGEF and PH domain containing 2; FYVE, RhoGEF and PH domain-containing protein 2; Tcd2; Tcd-2; t-complex distorter 2; tcs2; tcs-2; ZFYVE4; Zinc finger FYVE domain-containing protein 4
Gene Symbols FGD2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 508-537 amino acids from the C-terminal region of human FGD2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221472, 26382
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.