Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FGD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580627
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580627 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580627 Supplier Invitrogen™ Supplier No. PA580627
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has specificity for Human FGD2.

Activates CDC42, a member of the Ras-like family of Rho- and Rac proteins, by exchanging bound GDP for free GTP. Activates JNK1 via CDC42 but not RAC1. Binds to phosphatidylinositol 4,5- bisphosphate, phosphatidylinositol 3,4,5-trisphosphate, phosphatidylinositol 5-monophosphate, phosphatidylinositol 4- monophosphate and phosphatidylinositol 3-monophosphate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGD2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene FGD2
Gene Accession No. Q7Z6J4
Gene Alias faciogenital dysplasia homolog 2; faciogenital dysplasia protein 2; Fgd1 family, member 2; Fgd2; Fgd2-like protein; FLJ00276 protein; FYVE, RhoGEF and PH domain containing 2; FYVE, RhoGEF and PH domain-containing protein 2; Tcd2; Tcd-2; t-complex distorter 2; tcs2; tcs-2; ZFYVE4; Zinc finger FYVE domain-containing protein 4
Gene Symbols FGD2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the center region of the Human FGD2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221472
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.