Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FGD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570564
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570564 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570564 Supplier Invitrogen™ Supplier No. PA570564
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 92%; Horse: 85%; Human: 100%; Mouse: 86%; Pig: 85%; Rabbit: 79%; Rat: 100%.

FGD3 gene ontology annotations related to this gene include regulation of Rho protein signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGD3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FGD3
Gene Accession No. Q5JSP0
Gene Alias 5830461L01Rik; faciogenital dysplasia 3; faciogenital dysplasia homolog 3; Fgd1 family, member 3; FGD3; FGD3 protein; FLJ00004; FYVE, RhoGEF and PH domain containing 3; FYVE, RhoGEF and PH domain-containing protein 3; ZFYVE5; Zinc finger FYVE domain-containing protein 5
Gene Symbols FGD3
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human FGD3 (aa 501-550).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89846
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.