Learn More
FGD3 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305198A

Description
The recommended shelf life for this product is 1 year from date of receipt.
FGD3 gene ontology annotations related to this gene include regulation of Rho protein signal transduction.Specifications
FGD3 | |
Polyclonal | |
Unconjugated | |
FGD3 | |
faciogenital dysplasia 3, FGD1 family, member 3, FLJ00004, ZFYVE5, zinc finger FYVE domain-containing protein 5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
89846 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q5JSP0 | |
FGD3 | |
Between 50 and 100 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.