Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FGD4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558706
Description
Immunogen sequence: EPLLDTHIVN GERDETATAP ASPTTDSCDG NASDSSYRTP GIGPVLPLEE RGAETETKVQ ERENGESPLE LEQLDQHHEM KETNEQ Highest antigen sequence identity to the following orthologs: Mouse - 53%, Rat - 52%.

Specifications
FGD4 | |
Polyclonal | |
Unconjugated | |
FGD4 | |
Actin filament-binding protein frabin; actin-filament binding protein frabin; CMT4H; FGD1 family, member 4; FGD1-related F-actin-binding protein; FGD4; FRABP; FYVE, RhoGEF and PH domain containing 4; FYVE, RhoGEF and PH domain-containing protein 4; ZFYVE6; Zinc finger FYVE domain-containing protein 6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
121512 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96M96 | |
FGD4 | |
Recombinant protein corresponding to Human FGD4. Recombinant protein control fragment (Product #RP-97793). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction