Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FGD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA522062
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522062 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522062 Supplier Invitrogen™ Supplier No. PA522062
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The FGD4 gene, also known as FYVE, RhoGEF, and PH domain-containing protein 4, is located on chromosome 12p11.21. This gene encodes a protein involved in the regulation of the actin cytoskeleton and is crucial for cellular processes such as migration and adhesion. FGD4 is specifically associated with Charcot-Marie-Tooth disease type 4H (CMT4H), a rare autosomal recessive hereditary neuropathy characterized by early onset, progressive distal muscle weakness, scoliosis, and myelin outfoldings. Mutations in FGD4 result in disruptions of its function, leading to the clinical manifestations of CMT4H. Additionally, the gene has been implicated in cancer biology, with its expression correlating with aggressive phenotypes in some cancers and contributing to therapeutic resistances. The gene includes multiple exons and alternative splicing variants, underlining its broad functional implications.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGD4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene FGD4
Gene Accession No. Q96M96
Gene Alias Actin filament-binding protein frabin; actin-filament binding protein frabin; CMT4H; FGD1 family, member 4; FGD1-related F-actin-binding protein; FGD4; FRABP; FYVE, RhoGEF and PH domain containing 4; FYVE, RhoGEF and PH domain-containing protein 4; ZFYVE6; Zinc finger FYVE domain-containing protein 6
Gene Symbols FGD4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human FGD4. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121512
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.