Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FGD6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552494
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552494 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552494 Supplier Invitrogen™ Supplier No. PA552494
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALKHYLLKPV QRIPQYRLLL TDYLKNLIED AGDYRDTQDA LAVVIEVANH ANDTMKQGDN FQKLMQIQYS LNGHHEIVQP GRVFLKEGIL MKLSRKVMQP RMFFLFNDAL LYTTPVQSGM YKLNNMLSLA GMKVRKPTQE A Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

May activate CDC42, a member of the Ras-like family of Rho- and Rac proteins, by exchanging bound GDP for free GTP. May play a role in regulating the actin cytoskeleton and cell shape.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGD6
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FGD6
Gene Accession No. Q6ZV73
Gene Alias FGD6; FYVE, RhoGEF and PH domain containing 6; FYVE, RhoGEF and PH domain-containing protein 6; KIAA1362; ZFYVE24; Zinc finger FYVE domain-containing protein 24
Gene Symbols FGD6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FGD6. Recombinant protein control fragment (Product #RP-89561).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55785
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.