Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGD6 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548123
Description
May activate CDC42, a member of the Ras-like family of Rho- and Rac proteins, by exchanging bound GDP for free GTP. May play a role in regulating the actin cytoskeleton and cell shape.
Specifications
FGD6 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q6ZV73 | |
FGD6 | |
KLH conjugated synthetic peptide between 1324-1352 amino acids from the C-terminal region of human FGD6. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
FGD6 | |
FYVE, RhoGEF and PH domain-containing protein 6, Zinc finger FYVE domain-containing protein 24, FGD6, KIAA1362, ZFYVE24 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55785 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction