Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-10 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299396V
Description
FGF-10 Polyclonal specifically detects FGF-10 in Human samples. It is validated for ELISA.Specifications
FGF-10 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
DyLight 405 | |
FGF-10, fibroblast growth factor 10, Keratinocyte growth factor 2, produced by fibroblasts of urinary bladder lamina propria | |
Produced in rabbits immunized with purified, recombinant FGF-10 (NP_004456.1; Gln38-Ser208). | |
0.1 mL | |
Angiogenesis, Biologically Active Proteins, Cell Cycle and Replication | |
2255 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction