Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-12 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $728.30
Specifications
| Antigen | FGF-12 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
FGF-12 Polyclonal specifically detects FGF-12 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| FGF-12 | |
| Polyclonal | |
| Rabbit | |
| Angiogenesis | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 2257 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:EPSLHEIGEKQGRSRKSSGTPTMNGGKVVNQDS | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| FGF-12, FGF12B, FHF-1, FHF1Fibroblast growth factor homologous factor 1, fibroblast growth factor 12, fibroblast growth factor 12B, fibroblast growth factor FGF-12b, Myocyte-activating factor | |
| FGF12 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title