Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FGF-13 1B Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1909SP
This item is not returnable.
View return policy
Description
FGF-13 Monoclonal specifically detects FGF-13 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
FGF-13 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
NP_378668 | |
FGF13 | |
E. coli-derived recombinant human FGF-13 isoform 1B Ala2-Thr192 Accession # NP_378668 | |
25 μg | |
Primary | |
Detects human FGF-13 1B in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) FGF-3, -4, -5, -6, -7, -9, -10, -11, -12, -16, -17, -19, -20, -21, rhFGF basic, recombinant mouse FGF-8b, or -8c is observed. | |
Human | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
298923 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FGF13, FGF-13, FGF2Fibroblast growth factor homologous factor 2, FHF2FHF-2, fibroblast growth factor 13 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2258 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction