Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-13 Antibody (298923), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1909K
Description
FGF-13 Monoclonal antibody specifically detects FGF-13 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
FGF-13 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
298923 | |
Western Blot, Flow Cytometry, CyTOF-ready | |
FGF-13, FGF2Fibroblast growth factor homologous factor 2, FHF2FHF-2, fibroblast growth factor 13 | |
E. coli-derived recombinant human FGF-13 isoform 1B, Ala2-Thr192, Accession # NP_378668 | |
0.1 mL | |
Angiogenesis, Biologically Active Proteins, Neuroscience | |
2258 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction