Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-13 Antibody (298923), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1909MFV610
Description
FGF-13 Monoclonal antibody specifically detects FGF-13 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
| FGF-13 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Angiogenesis, Biologically Active Proteins, Neuroscience | |
| 2258 | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry, CyTOF | |
| 298923 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| FGF-13, FGF2Fibroblast growth factor homologous factor 2, FHF2FHF-2, fibroblast growth factor 13 | |
| E. coli-derived recombinant human FGF-13 isoform 1B, Ala2-Thr192, Accession # NP_378668 | |
| RUO | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction