Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-13 Antibody (OTI7E8), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270721F
Description
FGF-13 Monoclonal specifically detects FGF-13 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
FGF-13 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
FGF13 | |
Full length recombinant protein of human FGF-13 (NP_004105) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Immunohistochemistry | |
OTI7E8 | |
Immunohistochemistry | |
FGF-13, FGF2Fibroblast growth factor homologous factor 2, FHF2FHF-2, fibroblast growth factor 13 | |
Mouse | |
Affinity Purified | |
Angiogenesis, Biologically Active Proteins, Neuroscience | |
2258.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction