Learn More
FGF-13 Antibody (S235-22), DyLight 755, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP242178IR
Catalog No. NBP242178IR
Only null left
Add to Cart
Description
Description
FGF-13 Monoclonal specifically detects FGF-13 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
FGF-13
Monoclonal
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
FGF13
Synthetic peptide amino acids 2-18 (AAAIASSLIRQKRQARE) of human FHF2A. 100% identical to rat, 94% identical to mouse. >80% identity with FGF12A/FHF1A, FGF14A/FHF4A and FGF11A/FHF3A.
0.1 mL
Primary
Greater then 80% identity with FGF12A/FHF1A, FGF14A/FHF4A and FGF11A/FHF3A. Detects approx 30kDa. Does not cross-react with FGF13B/FHF2B. Cross reacts with FGF12A/FHF1A and FGF14A/FHF4A.
Store at 4C in the dark.
IgG2b
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
S235-22
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
FGF-13, FGF2Fibroblast growth factor homologous factor 2, FHF2FHF-2, fibroblast growth factor 13
Mouse
Protein G purified
Angiogenesis, Biologically Active Proteins, Neuroscience
2258
Human, Mouse, Rat
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.