Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-13 Antibody (S235-22), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242178H
Description
FGF-13 Monoclonal specifically detects FGF-13 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
FGF-13 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
FGF13 | |
Synthetic peptide amino acids 2-18 (AAAIASSLIRQKRQARE) of human FHF2A. 100% identical to rat, 94% identical to mouse. >80% identity with FGF12A/FHF1A, FGF14A/FHF4A and FGF11A/FHF3A. | |
0.1 mL | |
Primary | |
Greater then 80% identity with FGF12A/FHF1A, FGF14A/FHF4A and FGF11A/FHF3A. Detects approx 30kDa. Does not cross-react with FGF13B/FHF2B. Cross reacts with FGF12A/FHF1A and FGF14A/FHF4A. | |
Store at 4C in the dark. | |
IgG2b |
Western Blot, Immunohistochemistry | |
S235-22 | |
Western Blot, Immunohistochemistry | |
FGF-13, FGF2Fibroblast growth factor homologous factor 2, FHF2FHF-2, fibroblast growth factor 13 | |
Mouse | |
Protein G purified | |
Angiogenesis, Biologically Active Proteins, Neuroscience | |
2258 | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction