Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-16 Antibody (MM0283-6G2), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212297AF532
Description
FGF-16 Monoclonal specifically detects FGF-16 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FGF-16 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
FGF16 | |
Human recombinant FGF-16 | |
Primary | |
Human | |
IgG2 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0283-6G2 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
FGF-16, fibroblast growth factor 16 | |
Mouse | |
0.1 mL | |
8823 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction