Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-17 Antibody (07), DyLight 594 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP305818DL594
Description
FGF-17 Monoclonal antibody specifically detects FGF-17 in Human samples. It is validated for ELISASpecifications
FGF-17 | |
Monoclonal | |
DyLight 594 | |
FGF-13, FGF-17, fibroblast growth factor 17 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human FGF-17 (O60258-1; Thr23-Thr216). | |
0.1 mL | |
Angiogenesis, Neuroscience | |
8822 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
07 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction