Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-19 Antibody (032), Alexa Fluor™ 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290078AF350
Description
FGF-19 Monoclonal specifically detects FGF-19 in Human samples. It is validated for ELISA.Specifications
FGF-19 | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human FGF6 Human FGF9 Human FGF10 Human FGF17 Human FGF18 Human FGF acidic Human FGF basic | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
32 | |
ELISA | |
FGF-19, fibroblast growth factor 19 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human FGF-19 (O95750; Phe27-Lys216). | |
0.1 mL | |
Angiogenesis, Biologically Active Proteins, Neuroscience | |
9965 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction