Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-19 Antibody (032), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290078APC
Description
FGF-19 Monoclonal antibody specifically detects FGF-19 in Human samples. It is validated for ELISASpecifications
FGF-19 | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
032 | |
ELISA | |
FGF-19, fibroblast growth factor 19 | |
Rabbit immunized with purified, recombinant Human FGF-19 (Uniprot#: O95750-1, Phe27-Lys216) | |
0.1 mL | |
Angiogenesis, Biologically Active Proteins, Neuroscience | |
9965 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction