Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-19 Antibody (11H3), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | FGF-19 |
---|---|
Clone | 11H3 |
Dilution | Western Blot 1:500-1:2000, ELISA |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
FGF-19 Monoclonal antibody specifically detects FGF-19 in Human samples. It is validated for Western Blot, ELISASpecifications
FGF-19 | |
Western Blot 1:500-1:2000, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
FGF-19, fibroblast growth factor 19 | |
A synthesized peptide derived from Human FGF-19 [UniProt O95750] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
11H3 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Angiogenesis, Biologically Active Proteins, Neuroscience | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
9965 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title