Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-21 Antibody (005), DyLight 755

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290579IR
Description
FGF-21 Monoclonal specifically detects FGF-21 in Mouse samples. It is validated for ELISA.Specifications
FGF-21 | |
Monoclonal | |
DyLight 755 | |
FGF-21, fibroblast growth factor 21 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse FGF-21 (Q9JJN1; Met1-Ser210). | |
0.1 mL | |
Angiogenesis, Biologically Active Proteins, Cancer | |
26291 | |
Mouse | |
Purified |
ELISA | |
5 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Has cross-reactivity in ELISA with: Human FGF21 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction