Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGF-21 Antibody (461804), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB25372S
Description
FGF-21 Monoclonal antibody specifically detects FGF-21 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
FGF-21 | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, CyTOF | |
461804 | |
Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
FGF-21, fibroblast growth factor 21 | |
E. coli-derived recombinant human FGF-21, His29-Ser209, Accession # Q9NSA1 | |
0.1 mL | |
Angiogenesis, Biologically Active Proteins, Cancer | |
26291 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction